Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21019

Sigma-Aldrich

Cesium sulfate

puriss. p.a., ≥99.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs2O4S
CAS Number:
Molecular Weight:
361.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.5% (T)
form:
solid
solubility:
H2O: 0.2 g/mL, clear, colorless

grade

puriss. p.a.

assay

≥99.5% (T)

form

solid

impurities

≤0.001% total nitrogen (N)

mp

1019 °C (lit.)

solubility

H2O: 0.2 g/mL, clear, colorless

density

4.243 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Cs+].[Cs+].[O-]S([O-])(=O)=O

InChI

1S/2Cs.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

FLJPGEWQYJVDPF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Kufe et al.
The Journal of biological chemistry, 256(17), 8885-8888 (1981-09-10)
We have employed cesium sulfate density gradient centrifugation to separate RNA and DNA of L1210 cells labeled with [3H]fluorodeoxyuridine. We have analyzed nucleotide and nucleoside digests of purified DNA from the [3H]fluorodeoxyuridine-labeled cells and demonstrate by reverse phase and anion
A K Attri et al.
Analytical biochemistry, 159(1), 88-95 (1986-11-15)
We present a technique for analytical buoyant density sedimentation of nucleic acids which is performed in a preparative ultracentrifuge, in contrast to an analytical ultracentrifuge. Following centrifugation in a preparative rotor, small cylindrical quartz tubes are optically scanned; upon completion
A Miyakawa et al.
Neuroscience, 134(2), 595-604 (2005-06-25)
Effects of neuropeptide Y (NPY) on substantia gelatinosa neurons were investigated in adult rat spinal cord slices using blind whole-cell patch-clamp technique. Bath application of NPY (1 microM) induced a membrane hyperpolarization, resulting in a suppression of the dorsal root
F A Aly et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 17(11), 1257-1261 (2002-01-05)
A flow-injection (FI) methodology using tris(2,2'-dipyridyl)ruthenium(II), [Ru(dipy)3(2+)], chemiluminescence (CL) was developed for the rapid and sensitive determination of three thioxanthene derivatives, namely zuclopenthixol hydrochloride, flupentixol hydrochloride and thiothixene. The method is based on the CL reaction of the studied thioxanthenes
D W Gruenwedel et al.
Biopolymers, 7(4), 557-570 (1969-01-01)
When DNA's of differing GC:AT base ratios, e.g. synthetic poly dAT, T4 DNA,calf thymus DNA, E. coli DNA, and M. lysodeikticus DNA, are heat-denatured at neutral pH in increasing concentrations of N(a)(2)SO(4) or C(s)(2)SO(4) as supporting electrolytes,the variation of melting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service