Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200794

Sigma-Aldrich

Cesium acetate

technical grade, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCs
CAS Number:
Molecular Weight:
191.95
Beilstein/REAXYS Number:
3595637
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
assay:
≥95%
grade:
technical grade
form:
solid

grade

technical grade

assay

≥95%

form

solid

mp

194 °C (lit.)

SMILES string

[Cs+].CC([O-])=O

InChI

1S/C2H4O2.Cs/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

ZOAIGCHJWKDIPJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium acetate is an inorganic salt. It has been reported to change the surface of kaolinite by undergoing intercalation under various reaction conditions.[1] It modifies the catalytic activity of MCM (Mobil Composition of Matter) -41 and MCM-48-type mesoporous molecular sieves by undergoing ion exchange with cesium cations and impregnation on their surfaces.[2]

Application

Cesium acetate may be used for the inversion of secondary alcohols to afford inverted acetates.[3] It can also be used for the inversion of cyclopentyl and cyclohexyl alcohols.[4]

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient inversions of secondary alcohols using cesium acetate and 18-crown-6.
Torisawa Y, et al.
Chemistry Letters (Jpn), 9, 1555-1556 (1984)
A procedure for alcohol inversion using cesium acetate.
Huffman JW and Desai RC.
Synthetic Communications, 13(7), 553-557 (1983)
Cesium-modified mesoporous molecular sieves as basic catalysts for Knoevenagel condensations.
Ernst S, et al.
Studies in Surface Science and Catalysis, 125, 367-374 (1999)
Modification of kaolinite surfaces with cesium acetate at 25, 120, and 220 C.
Frost RL, et al.
Langmuir, 15(26), 8787-8794 (1999)
Efficient method for inversion of secondary alcohols by reaction of chloromethanesulfonates with cesium acetate.
Shimizu T, et al.
Tetrahedron Letters, 37(34), 6145-6148 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service