Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15513

Sigma-Aldrich

Formaldehyde solution

meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein/REAXYS Number:
1209228
MDL number:
UNSPSC Code:
12352114
eCl@ss:
39021101
PubChem Substance ID:
bp:
97 °C
vapor pressure:
52 mmHg ( 37 °C)

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

quality

meets analytical specification of Ph. Eur., BP

contains

10% methanol as stabilizer

concentration

35 wt. %
35.0-38.0%

impurities

acidic reac. substances, complies
residual solvents, complies
9.0-15.0% methanol (GC)

ign. residue

≤0.01% (as SO4)

bp

97 °C

mp

-15 °C

density

1.09 g/mL at 25 °C (lit.)

suitability

complies for appearance of solution

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

Inchi Key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

133.0 °F - closed cup

flash_point_c

56.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy S Ma et al.
The Journal of experimental medicine, 205(7), 1551-1557 (2008-07-02)
Hyper-immunoglobulin E syndrome (HIES) is a primary immune deficiency characterized by abnormal and devastating susceptibility to a narrow spectrum of infections, most commonly Staphylococcus aureus and Candida albicans. Recent investigations have identified mutations in STAT3 in the majority of HIES
G R Moore et al.
Neurology, 55(10), 1506-1510 (2000-11-30)
To determine the pathologic basis of areas not exhibiting signal of the short-T2 component of the T2 relaxation distribution in MS, as studied in formalin-fixed brain. A myelin-specific MRI signal would be of great importance in assessing demyelination in patients
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
Richard Brad Jones et al.
PLoS pathogens, 10(8), e1004287-e1004287 (2014-08-15)
Resting memory CD4+ T-cells harboring latent HIV proviruses represent a critical barrier to viral eradication. Histone deacetylase inhibitors (HDACis), such as suberanilohydroxamic acid (SAHA), romidepsin, and panobinostat have been shown to induce HIV expression in these resting cells. Recently, it
Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service