Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154814

Sigma-Aldrich

N,N-Dimethylformamide

ACS spectrophotometric grade, ≥99.8%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein/REAXYS Number:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39031202
PubChem Substance ID:
grade:
ACS spectrophotometric grade
assay:
≥99.8%
technique(s):
UV/Vis spectroscopy: suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

grade

ACS spectrophotometric grade

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

assay

≥99.8%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0005 meq/g Titr. acid
≤0.003 meq/g Titr. base
≤0.15% water

evapn. residue

≤0.005%

color

APHA: ≤15
clear

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

λ

H2O reference

UV absorption

λ: 270 nm Amax: 1.00
λ: 275 nm Amax: 0.30
λ: 295 nm Amax: 0.10
λ: 310 nm Amax: 0.05
λ: 340-400 nm Amax: 0.01

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Solvent for many hydrophobic organic compounds.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

135.5 °F - closed cup

flash_point_c

57.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Liu et al.
Nanoscale, 4(20), 6574-6580 (2012-09-15)
Studies on the self-assembly of metal nanoparticles (NPs) in the presence of ions are motivated by the biosensing applications of NP clusters and the capability to control the morphology of clusters of oppositely charged NPs. The effect of ions has
L Piazza et al.
Nature communications, 6, 6407-6407 (2015-03-03)
Surface plasmon polaritons can confine electromagnetic fields in subwavelength spaces and are of interest for photonics, optical data storage devices and biosensing applications. In analogy to photons, they exhibit wave-particle duality, whose different aspects have recently been observed in separate
Jun Nishida et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(52), 18442-18447 (2014-12-17)
The structural elasticity of metal-organic frameworks (MOFs) is a key property for their functionality. Here, we show that 2D IR spectroscopy with pulse-shaping techniques can probe the ultrafast structural fluctuations of MOFs. 2D IR data, obtained from a vibrational probe
S Ollier et al.
Journal of dairy science, 97(8), 4942-4954 (2014-06-03)
A cow's risk of acquiring a new intramammary infection during the dry period increases with milk production at drying off and decreases as mammary gland involution progresses. A method commonly used to reduce milk production is a drastic reduction in
Guillaume E Beranger et al.
Endocrinology, 155(4), 1340-1352 (2014-02-11)
Osteoporosis and overweight/obesity constitute major worldwide public health burdens that are associated with aging. A high proportion of women develop osteoporosis and increased intraabdominal adiposity after menopause. which leads to bone fractures and metabolic disorders. There is no efficient treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service