Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15370

Sigma-Aldrich

Lead tetraacetate

purum p.a., moistened with ~5% acetic acid to stabilize, ≥95% (dried material)

Synonym(s):

Lead(IV) acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3COO)4
CAS Number:
Molecular Weight:
443.38
Beilstein/REAXYS Number:
3595640
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
assay:
≥95% (dried material)
form:
solid

grade

purum p.a.

assay

≥95% (dried material)

form

solid

quality

moistened with ~5% acetic acid to stabilize

SMILES string

CC(=O)O[Pb](OC(C)=O)(OC(C)=O)OC(C)=O

InChI

1S/4C2H4O2.Pb/c4*1-2(3)4;/h4*1H3,(H,3,4);/q;;;;+4/p-4

InChI key

JEHCHYAKAXDFKV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

For the preparation of a dip reagent for the fluorimetric determination of diols, glycosides and phenols (see also 35848 2′,7′-Dichlorofluorescein)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Chang et al.
Journal of cellular biochemistry, 120(9), 15369-15377 (2019-05-01)
Long noncoding RNAs (lncRNAs) have been implicated in colorectal cancer (CRC). And lncRNA RP11-138J23.1 (CRCAL-3) was previously reported as a candidate regulator of CRC development. But its regulating functions have not been fully elucidated. Here, we analyzed RNA sequencing data
Huayou Hu et al.
Journal of the American Chemical Society, 131(33), 11998-12006 (2009-08-06)
A series of seven cyclopent-3-en-1-ylmethylamines bearing one, two, or three methyl substituents at the C2, C3, C4, or C(alpha) positions, including the unsubstituted parent, was accessed by ring-closing metatheses of alpha,alpha-diallylacetonitrile (or methallyl variants) and alpha,alpha-diallylacetone followed by hydride reductions
Glycol-cleavage oxidation.
Arthur S Perlin
Advances in carbohydrate chemistry and biochemistry, 60, 183-250 (2006-06-06)
T H Ma et al.
Mutation research, 270(1), 71-77 (1992-11-01)
Synergistic and antagonistic effects on genotoxicity of mixtures of four chemicals; i.e., lead tetraacetate (LTA), arsenic trioxide (ATO), dieldrin (DED), and tetrachloroethylene (TCE), were evaluated by the Tradescantia-micronucleus (Trad-MCN) assay. The chemicals were mixed in ratios of 1:1, 1:2 and
Luis Moreno-Osorio et al.
Magnetic resonance in chemistry : MRC, 45(11), 993-996 (2007-09-19)
Two naturally occurring 11-nordrimanes were synthesized, and their (1)H and (13)C NMR spectra were unambiguously assigned in full for the first time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service