Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14527

Sigma-Aldrich

Tin(II) oxide

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnO
CAS Number:
Molecular Weight:
134.71
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥95%

reaction suitability

reagent type: catalyst
core: tin

impurities

≤5% SnO2

SMILES string

O=[SnH2]

InChI

1S/O.Sn

InChI key

QHGNHLZPVBIIPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Römpp et al.
Analytical and bioanalytical chemistry, 407(8), 2329-2335 (2015-01-13)
Mass spectrometry imaging has become a popular tool for probing the chemical complexity of biological surfaces. This led to the development of a wide range of instrumentation and preparation protocols. It is thus desirable to evaluate and compare the data
Sean P Hill et al.
The journal of physical chemistry letters, 6(22), 4510-4517 (2015-11-05)
Molecular photon upconversion via triplet-triplet annihilation (TTA-UC), combining two or more low energy photons to generate a higher energy excited state, is an intriguing strategy to surpass the maximum efficiency for a single junction solar cell (<34%). Here, we introduce
Zheng Tian et al.
Langmuir : the ACS journal of surfaces and colloids, 30(41), 12411-12420 (2014-09-25)
Carbon films with interdigitated bimodal three-dimensionally ordered mesoporosity (ib3DOm) are realized by a scalable nanoreplication process that removes the common need plaguing hard-templating strategies for multistep prefabrication of porous sacrificial templates. Specifically, evaporation-induced convective codeposition of size-tunable (ca. 20-50 nm)
Sangheon Lee et al.
Scientific reports, 5, 17472-17472 (2015-12-08)
Semiconductor sensitized solar cells, a promising candidate for next-generation photovoltaics, have seen notable progress using 0-D quantum dots as light harvesting materials. Integration of higher-dimensional nanostructures and their multi-composition variants into sensitized solar cells is, however, still not fully investigated
Lukas Krasny et al.
Analytical and bioanalytical chemistry, 407(8), 2141-2147 (2014-11-05)
Nanostructure-assisted laser desorption/ionization (NALDI) has been recognized as a powerful matrix-free mass spectrometry tool ideal for imaging of small molecules. In this report, the NALDI approach was compared with matrix-assisted laser desorption/ionization (MALDI) mass spectrometry in terms of sensitivity, reproducibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service