Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13914

Sigma-Aldrich

Strontium nitrate

puriss., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(NO3)2
CAS Number:
Molecular Weight:
211.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥98%
form:
solid

grade

puriss.

assay

≥98%

form

solid

impurities

≤0.001% heavy metals (as Pb)

loss

≤0.5% loss on drying, 105 °C

pH

5-7 (20 °C, 5%)

mp

570 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤200 mg/kg
Fe: ≤5 mg/kg
Mg: ≤20 mg/kg
Na: ≤500 mg/kg

SMILES string

[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Sr/c2*2-1(3)4;/q2*-1;+2

InChI key

DHEQXMRUPNDRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strontium nitrate was used in the preparation of La0.4Sr0.5Ba0.1TiO3 (LSBT) precursor solution.[1]

pictograms

Flame over circleCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhai et al.
Contact dermatitis, 42(2), 98-100 (2000-03-07)
Skin care products are complex formulations that may cause sensory irritation symptoms, characterized by stinging, burning, and itching. Substances capable of counteracting sensory irritation are of great practical interest. Strontium salts have been demonstrated to inhibit sensory irritation and inflammation
Sangita Pandey et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 220-226 (2011-02-08)
Electron paramagnetic resonance (EPR) study of Fe³(+) ions doped strontium nitrate (SN) single crystals is performed at liquid nitrogen temperature and at X band frequency. The spin Hamiltonian (SH) parameters are determined from the resonance lines observed at different angular
H Zhai et al.
Dermatology (Basel, Switzerland), 200(3), 244-246 (2000-06-01)
Previous studies have demonstrated that strontium salts have a potent and broad ability to suppress sensory irritation. To ascertain the possible antipruritic effects of topical strontium salts, we conducted a double-blind randomized study to evaluate the effect of 20% strontium
Structure refinement of strontium nitrate, Sr(NO3)2, and barium nitrate, Ba(NO3)2.
Nowotny H and Heger G.
Acta Crystallographica Section C, Crystal Structure Communications, 39(8), 952-956 (1983)
G S Hahn
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 25(9), 689-694 (1999-09-22)
Irritant contact dermatitis occurs following topical exposure to many chemicals found in cosmetics, personal care products, drugs, and during occupational exposure. Signs and symptoms may include sensory irritation (sting, burn, and/or itch), erythema, edema, and vesiculation. In an attempt to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service