Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13729

Sigma-Aldrich

Sodium silicate solution

purum, ≥10% NaOH basis, ≥27% SiO2 basis

Synonym(s):

Sodium trisilicate solution, Water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O(SiO2)x · xH2O
UNSPSC Code:
12352202

grade

purum

composition

NaOH, ≥10%
SiO2, ≥27%

density

1.39 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of layered silicates from sodium silicate solution.
Kwon O-Y and Park K-W.
Bull. Korean Chem. Soc., 25(1), 25-26 (2004)
Chemical optimisation of the compressive strength of aluminosilicate geopolymers synthesised by sodium silicate activation of metakaolinite.
Rowles M and O'connor B.
Journal of Materials Chemistry, 13(5), 1161-1165 (2003)
F Miyaji et al.
Biomaterials, 20(10), 913-919 (1999-06-03)
A bonelike apatite layer was formed on organic polymers when sodium silicate was used as a catalyst for the apatite nucleation, and modified simulated body fluid was used as a medium for the apatite growth. The apatite-forming ability was the
Synthesis of mesoporous silica with tunable pore size from sodium silicate solutions and a polyethylene oxide surfactant.
Sierra L and Guth J-L.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 27(2), 243-253 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service