Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13726

Sigma-Aldrich

Sodium trisilicate

≥18% Na (as Na2O) basis, ≥60% Si (as SiO2) basis, powder

Synonym(s):

Sodium water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O · 3SiO2
CAS Number:
Molecular Weight:
242.23
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

composition

Na (as Na2O), ≥18%
Si (as SiO2), ≥60%

loss

≤20% loss on ignition, 800 °C

SMILES string

[Na+].[Na+].[O-][Si]([O-])=O

InChI

1S/2Na.O3Si/c;;1-4(2)3/q2*+1;-2

InChI key

NTHWMYGWWRZVTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interaction of ultraviolet excimer laser light with sodium trisilicate.
Eschbach PA, et al.
J. Vac. Sci. Technol. B, 7(5), 2943-2951 (1989)
Yoon-Ha Kim et al.
Biological trace element research, 144(1-3), 1175-1185 (2011-04-06)
The current study was conducted in order to investigate the short-term effects (6, 12, and 24 h) of silicon (Si) on the endogenous hormonal composition of rice (Oryza sativa L. cv. Dongjin-beyo), with and without wounding stress. Si applied in different
Monique Tohoué Tognonvi et al.
Journal of colloid and interface science, 352(2), 309-315 (2010-10-01)
Both concentrated and diluted sodium silicate solutions have been investigated by combining (29)Si NMR spectroscopy and SAXS experiments. The chemical nature of the entities responsible for the high siliceous species solubility observed in such alkaline concentrated sodium silicate solutions and
Karl Mandel et al.
ACS applied materials & interfaces, 4(10), 5633-5642 (2012-09-14)
The synthesis of a reusable, magnetically switchable nanocomposite microparticle, which can be modified to selectively extract and recover Hg(II) or Cu(II) from water, is reported. Superparamagnetic iron oxide (magnetite) nanoparticles act as the magnetic component in this system, and these
Karin E Sisti et al.
The Journal of oral implantology, 38(3), 231-237 (2010-08-10)
Surface and biomechanical analysis of titanium implant surfaces modified by laser beam with and without hydroxyapatite. Titanium implants with 3 different surfaces were inserted into the tibias of 30 rabbits: group I (GI) machined surface (control group), group II irradiated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service