Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13117

Sigma-Aldrich

Magnesium carbonate basic

puriss., meets analytical specification of Ph. Eur., BP, E504, light, 40-45% Mg (as MgO) basis, powder (light)

Synonym(s):

Magnesium hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCO3
CAS Number:
Molecular Weight:
84.31
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21
form:
powder (light)

grade

puriss.

form

powder (light)

quality

meets analytical specification of Ph. Eur., BP, E504

composition

Mg (as MgO), 40-45%

impurities

residual solvents, complies
≤0.001% heavy mteals (as Pb)
≤0.05% insoluble in acetic acid
≤1% in water soluble matter

anion traces

chloride (Cl-): ≤300 mg/kg
sulfate (SO42-): ≤2000 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤4000 mg/kg
Cu: ≤10 mg/kg
Fe: ≤200 mg/kg
Hg: ≤1 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

[Mg++].O[Mg]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

UDYRPJABTMRVCX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium carbonate basic (Magnesium hydroxy carbonate) has been synthesized by reacting magnesium hydroxide and carbon dioxide.[1]

Application

Magnesium carbonate basic may be employed for the isolation of carotenoids from grapes during the quantification of carotenoids in grapes by reversed-phase C30 (RP-30) HPLC-DAD-MS (ESI+) (High-performance liquid chromatography-diode array detection-mass spectrometry with electrospray ionization) analysis.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of a magnesium hydroxy carbonate from magnesium hydroxide.
Botha A and Strydom CA.
Hydrometallurgy, 62(1), 175-183 (2001)
Pasquale Crupi et al.
Journal of mass spectrometry : JMS, 45(9), 971-980 (2010-09-04)
Carotenoids are uniquely functional polyene pigments ubiquitous in nature; aside from being responsible for the color of a wide variety of vegetables, interest is being focused on food carotenoids due to their likely health benefits. From analytical point of view

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service