Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13029

Sigma-Aldrich

Lithium sulfate monohydrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4 · H2O
CAS Number:
Molecular Weight:
127.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥98%
form:
solid

assay

≥98%

form

solid

impurities

≤0.001% heavy metals (as Pb)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤20 mg/kg
K: ≤200 mg/kg
Na: ≤100 mg/kg

SMILES string

[Li+].[Li+].O.[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

RKGLUDFWIKNKMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lithium sulfate monohydrate is a colorless crystalline lithium salt. Its solubility in water decreases with a rise in temperature. It can be synthesized by the dissolution of lithium carbonate in sulfuric acid.[1] A kinetic model has been proposed to investigate the dehydration of Li2SO4.H2O. Monte-Carlo simulation of the various processes (nucleation and growth) involved in its dehydration has been evaluated.[2] Neutron-diffraction investigations of LiSO4.H2O have been reported.[3]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Schmidt et al.
Acta crystallographica. Section A, Foundations of crystallography, 65(Pt 4), 267-275 (2009-06-19)
Piezoelectric lithium sulfate monohydrate, Li2SO4.H2O, was analyzed with respect to the relationship between the static structural properties of the crystal and its response to an external electric field. The static electron density was determined via standard low-temperature X-ray data collection
Neutron-Diffraction Study of Lithium Sulfate Monohydrate.
Smith HG, et al.
J. Chem. Phys. , 48(12), 5561-5565 (1968)
Loïc Favergeon et al.
Physical chemistry chemical physics : PCCP, 7(21), 3723-3727 (2005-12-17)
A kinetic model for the dehydration of lithium sulfate monohydrate is proposed in order to account for experimental data obtained on single crystals by thermogravimetry at 80 degrees C under fixed water vapour pressure, and by optical microscopy. This model
Eagleson M.
Concise Encyclopedia Chemistry, 605-605 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service