Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13010

Sigma-Aldrich

Lithium carbonate

purum, ≥99%

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein/REAXYS Number:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

purum

assay

≥99%

form

solid

impurities

≤0.003% heavy metals (as Pb)

mp

618 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤3000 mg/kg

cation traces

Ca: ≤300 mg/kg
Fe: ≤30 mg/kg
K: ≤300 mg/kg
Mg: ≤100 mg/kg
Na: ≤1000 mg/kg

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanadium modified LiFePO4 cathode for Li-ion batteries.
Hong J, et al
Electrochemical and Solid-State Letters, 12(2), A33-A38 (2009)
Xiaoyue Tan et al.
Journal of the American Society of Nephrology : JASN, 17(12), 3382-3393 (2006-11-04)
Deficiency in vitamin D and its active metabolites is a pathologic feature of chronic kidney diseases. Despite that tubular epithelial cells are the major sites of active vitamin D synthesis, little is known about the role of vitamin D in
Bettina Ziegelmann et al.
Scientific reports, 8(1), 683-683 (2018-01-14)
Honey bees are increasingly important in the pollination of crops and wild plants. Recent reports of the weakening and periodical high losses of managed honey bee colonies have alarmed beekeeper, farmers and scientists. Infestations with the ectoparasitic mite Varroa destructor
Tom Bschor et al.
Journal of clinical psychopharmacology, 33(1), 38-44 (2013-01-02)
Studies of the 1970s and 1980s showed lithium monotherapy to be an effective treatment of acute unipolar major depressive disorder (MDD) and hence as a potential alternative to monoaminergic antidepressants.The objective was to conduct the first comparison of a lithium
Andrea Contestabile et al.
The Journal of clinical investigation, 123(1), 348-361 (2012-12-04)
Down syndrome (DS) patients exhibit abnormalities of hippocampal-dependent explicit memory, a feature that is replicated in relevant mouse models of the disease. Adult hippocampal neurogenesis, which is impaired in DS and other neuropsychiatric diseases, plays a key role in hippocampal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service