Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12605

Sigma-Aldrich

Potassium bisulfate

puriss., 98-102%

Synonym(s):

Potassium hydrogen sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHSO4
CAS Number:
Molecular Weight:
136.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
98-102%
form:
solid

grade

puriss.

assay

98-102%

form

solid

impurities

≤0.002% ammonium (NH4)
≤0.002% heavy metals (as Pb)

mp

214 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

Ca: ≤200 mg/kg
Fe: ≤20 mg/kg

SMILES string

[K+].OS([O-])(=O)=O

InChI

1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1

InChI key

CHKVPAROMQMJNQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viralkumar Patel et al.
The American journal of gastroenterology, 104(4), 953-965 (2009-02-26)
Ingestion of a concentrated low-volume phosphate solution produces copious diarrhea, which cleanses the colon, but it occasionally causes renal failure due to calcium phosphate precipitation in renal tubules. We hypothesized that a concentrated low-volume sulfate solution would be an equally
Douglas K Rex et al.
Gastrointestinal endoscopy, 72(2), 328-336 (2010-07-22)
Low-volume bowel preparations for colonoscopy improve tolerability. We compared the efficacy, tolerability, and safety of a new low-volume sulfate solution with a standard 4-L polyethylene glycol electrolyte lavage solution as bowel preparation for colonoscopy. Multicenter, single-blind, randomized, noninferiority study. Five
Tian-Li Ma et al.
BMC plant biology, 12, 161-161 (2012-09-12)
Potassium (K+) is an important nutrient ion in plant cells and plays crucial roles in many plant physiological and developmental processes. In the natural environment, K+ deficiency is a common abiotic stress that inhibits plant growth and reduces crop productivity.
Sunghee Lee et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 9556-9564 (2010-02-18)
In this work, we have investigated the effect of potassium salts of different anions upon the crystal nucleation of K(2)SO(4) as interfacially templated by a surfactant monolayer of 1-octadecylamine (ODA), in an aqueous microdroplet system bounded by a liquid-liquid interface
M G M Alam et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 42(4), 447-451 (2007-03-17)
This study presents an environmental-friendly and cost effective method for the extraction of arsenic from contaminated soils. Laboratory experiments using inorganic salts, potassium phosphate (KH2PO4), potassium chloride (KCl), potassium nitrate (KNO3), potassium sulfate (K2SO4), and sodium perchlorate (NaClO4) were evaluated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service