Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12344

Sigma-Aldrich

Iron(III) oxide

puriss., ≥97.0% (complexometric)

Synonym(s):

Ferric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Fe2O3
CAS Number:
Molecular Weight:
159.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥97.0% (complexometric)
form:
solid

grade

puriss.

assay

≥97.0% (complexometric)

form

solid

loss

≤1% loss on ignition

cation traces

Ca: ≤500 mg/kg
Cu: ≤100 mg/kg
Mg: ≤300 mg/kg
Zn: ≤200 mg/kg

SMILES string

O=[Fe]O[Fe]=O

InChI

1S/2Fe.3O

InChI key

JEIPFZHSYJVQDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The structure of iron (III) oxide-hydrate gels.
Van der Giessen AA.
J. Inorg. Nucl. Chem., 28(10), 2155-2159 (1966)
Synthesis and photoelectrochemical properties of nanoporous iron (III) oxide by potentiostatic anodization.
Prakasam HE, et al.
Nanotechnology, 17(17), 4285-4285 (2006)
William B Ingler et al.
Journal of the American Chemical Society, 126(33), 10238-10239 (2004-08-19)
Stable zinc-doped iron(III) oxide thin films that exhibit p-type behavior were synthesized by spray pyrolytic deposition (SPD) on conducting indium-doped tin oxide-coated glass substrate. The highest photocurrent density of 1.1 mA/cm2 was observed at an illumination intensity of 40 mW/cm2
The single-crystal Raman spectra of nearly opaque materials. Iron (III) oxide and chromium (III) oxide.
Beattie IR and Gilson TR.
J. Chem. Soc. Sect. A, 980-986 (1970)
Light-induced reduction of natural iron (III) oxide and its relevance to phytoplankton.
Finden DAS, et al.
Nature, 309, 783-784 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service