Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12304

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

puriss., ≥99% (manganometric)

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
assay:
≥99% (manganometric)
form:
solid

grade

puriss.

assay

≥99% (manganometric)

form

solid

shelf life

Shelf life is limited

reaction suitability

reagent type: catalyst
core: iron

pH

3-5 (20 °C, 50 g/L)

mp

100 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤200 mg/kg
Cu: ≤20 mg/kg
Fe3+: ≤500 mg/kg
K: ≤100 mg/kg
Mg: ≤200 mg/kg
Na: ≤100 mg/kg
Zn: ≤100 mg/kg

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Caution

will be oxidized by air

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of Tutton's salts. VI. Vanadium (II), iron (II) and cobalt (II) ammonium sulfate hexahydrates.
Montgomery H, et al.
Acta Crystallographica, 22(6), 775-780 (1967)
Gong Zhengjun et al.
Talanta, 66(4), 1012-1017 (2008-10-31)
A simple, accurate, sensitive and selective flow injection catalytic kinetic spectrophotometric method for rapid determination of trace amounts of selenium is proposed in this paper. The proposed method is based on the accelerating effect of Se(IV) on the reaction of
Mixed ligand complexes of iron (II) and (III) with cyanide and aromatic di-imines.
Schilt AA.
Journal of the American Chemical Society, 82(12), 3000-3005 (1960)
Yu-Jung Chang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 21(4), 520-524 (2005-04-07)
This study examined whether iron accumulated in ferritin-producing recombinant microbes is bioavailable to rats with iron deficiency. Rats induced with iron deficiency were treated with iron preparations of ferrous ammonium sulfate, horse spleen ferritin, control yeast, and ferritin-producing recombinant yeast
Y J Tseng et al.
Medical physics, 32(4), 1017-1023 (2005-05-18)
A least-squares error minimization approach was adopted to assess ferric ion diffusion coefficient of Fricke-agarose gels. Ferric ion diffusion process was modeled as a Gaussian-shaped degradation kernel operating on an initial concentration distribution. Diffusion coefficient was iteratively determined by minimizing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service