form
liquid
impurities
~0.5% benzene
refractive index
n20/D 1.388
bp
65-100 °C
density
0.692 g/mL at 20 °C
Looking for similar products? Visit Product Comparison Guide
Other Notes
Sales restrictions may apply
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
GC methods for quantitative determination of benzene in gasoline.
Pavlova A and Ivanova R.
Acta Chromatographica , 215-225 (2003)
Hosein Karim et al.
Journal of injury & violence research, 7(2), 59-63 (2015-06-18)
To investigate factors related to prevention of self-immolation in west of Iran. In a case-control study, 30 consecutive cases of deliberate self-inflicted burns admitted to the regional burn centre (Imam Khomeini hospital in Kermanshah province, Iran) were compared with controls
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service