Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11730

Sigma-Aldrich

Barium carbonate

purum p.a., ≥98.5% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCO3
CAS Number:
Molecular Weight:
197.34
Beilstein/REAXYS Number:
7045119
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

purum p.a.

assay

≥98.5% (KT)

anion traces

chloride (Cl-): ≤300 mg/kg

cation traces

Ca: ≤1000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤2000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Sr: ≤1.5%
Zn: ≤50 mg/kg

SMILES string

[Ba++].[O-]C([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Khorsand Zak et al.
Nanoscale research letters, 9(1), 399-399 (2014-09-02)
Pure zinc oxide and zinc oxide/barium carbonate nanoparticles (ZnO-NPs and ZB-NPs) were synthesized by the sol-gel method. The prepared powders were characterized by X-ray diffraction (XRD), ultraviolet-visible (UV-Vis), Auger spectroscopy, and transmission electron microscopy (TEM). The XRD result showed that
Anna V Faleva et al.
International journal of biological macromolecules, 166, 913-922 (2020-11-05)
In this article, the functional group composition of the spruce (Pícea ábies) and birch (Bétula péndula) phloem lignin is described. The features of the chemical structure were studied by analyzing dioxane lignin using the elemental analysis, UV-Vis, FT-IR, and 1D
Characterization of barium titanate powders: barium carbonate identification.
Lopez MCB, et al.
Journal of the American Chemical Society, 82(7), 1777-1786 (1999)
Jian-Hua Zhu et al.
Chemical communications (Cambridge, England), (9)(9), 1106-1108 (2009-02-20)
A facile biomimetic method is reported for the synthesis of novel BaCO(3) nanofibres with double-stranded and cylindrical helical morphologies via a phosphonated block co-polymer-controlled mineralization process.
D Ekeberg et al.
Carbohydrate research, 335(2), 141-146 (2001-09-25)
A simple, but low-yielding method for the synthesis of 3-hexuloses has been elaborated. Oxidation of 1,2:5,6-di-O-isopropylidenehexitols with bromine in the presence of barium carbonate, followed by mild-acid hydrolysis of the oxidation products gave the free hexuloses. Oxidation occurred at only

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service