Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11504

Sigma-Aldrich

Lead(II) acetate trihydrate

puriss., ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein/REAXYS Number:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.5%

grade

puriss.

assay

≥99.5%

form

solid

mp

75 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

Cu: ≤10 mg/kg
Fe: ≤10 mg/kg

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

target_organs

Central nervous system,Blood,Immune system,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbone L, et al.
Journal of Materials Chemistry, 16(40), 3952-3956 (2006)
Jane V Knichal et al.
Chemical communications (Cambridge, England), 50(92), 14436-14439 (2014-10-11)
5-Ethynyl-1,3-benzenedicarboxylic acid (H2ebdc) reacted with lead(II) acetate trihydrate yields a 1D ladder network, [Pb(ebdc)(MeOH)]2·H2O (1). Removing crystals of 1 from the mother liquor results in a facile single crystal to single crystal transition, yielding 2D [Pb(ebdc)] net (2) with a
Hazardous Chemicals: Safety Management and Global Regulations, 244-244 (2013)
Chunping Li et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 225-229 (2013-03-27)
Occupational and environmental exposures to lead (Pb) are a worldwide concern. DNA methylation plays an important role in the development of Pb toxicity. Here, we try to find out the evidence to prove that the methylation of the LINE-1 promoter
Mohammed H Badawoud et al.
Neurosciences (Riyadh, Saudi Arabia), 18(1), 52-57 (2013-01-08)
To investigate the morphological changes in the peripheral motor system of the rat induced by exposure to lead. This study was conducted at the Anatomy Department, Faculty of Medicine, King Abdulaziz University, Jeddah, Kingdom of Saudi Arabia from January 2011

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service