Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11428

Sigma-Aldrich

Barium peroxide

≥95% peroxide (as BaO2) basis, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaO2
CAS Number:
Molecular Weight:
169.33
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38110504
PubChem Substance ID:
assay:
≥95% peroxide (as BaO2) basis
form:
powder

assay

≥95% peroxide (as BaO2) basis

form

powder

reaction suitability

reagent type: oxidant

impurities

≤0.01% heavy metals (as Pb)
≤0.5% insoluble in HCl
≤1% substances not precipitated by H2SO4 (as SO4)

loss

≤1% loss on drying, 105 °C

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[Ba++].[O-][O-]

InChI

1S/Ba.O2/c;1-2/q+2;-2

InChI key

ZJRXSAYFZMGQFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincoli JW.
Risk Management for Hazardous Chemicals, 226-226 (1996)
On-line chemiluminescence detection for capillary electrophoresis based on the reaction of barium peroxide with luminescence reagents.
Lin J-M, et al.
Journal of Chromatography A, 844(1), 341-348 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service