Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10015

Sigma-Aldrich

Mercury(II) iodide red

puriss., red, ≥99%

Synonym(s):

Mercuric iodide red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgI2
CAS Number:
Molecular Weight:
454.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%

grade

puriss.

assay

≥99%

impurities

≤0.01% water-soluble heavy metals (as Pb)

ign. residue (after red. with HCOOH)

≤0.02% (as SO4)

SMILES string

I[Hg]I

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mokshada Jain et al.
PloS one, 16(1), e0243854-e0243854 (2021-01-14)
Family planning is a key means to achieving many of the Sustainable Development Goals. Around the world, governments and partners have prioritized investments to increase access to and uptake of family planning methods. In Uttar Pradesh, India, the government and
Diiodobis (1-methyl-1, 3-imidazolium-2-thiolato-S) mercury (II).
Pavlovic G, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 56(7), 801-803 (2000)
Eagleson M.
Concise Encyclopedia Chemistry, 637-637 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 542-542 (1994)
Concurrent mercuric blister and dimethyl sulphoxide (DMSO) application as a cause of mercury toxicity in two horses.
J C Schuh et al.
Equine veterinary journal, 20(1), 68-71 (1988-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service