Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06306

Sigma-Aldrich

Sodium hydroxide

≥98.5%, microprills

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

assay

≥98.5%

form

microprills

impurities

≤1% Na2CO3

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The aldol condensation.
Nielsen AT and Houlihan WJ.
Organic Reactions (1968)
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at
M Chevallet et al.
Electrophoresis, 19(11), 1901-1909 (1998-09-18)
Severe quantitative loss of protein is often observed in high-resolution two-dimensional electrophoresis of membrane proteins, while the resolution is usually not affected. To improve the solubility of proteins in this technique, we tested denaturing cocktails containing various detergents and chaotropes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service