Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04211

Sigma-Aldrich

Ammonium phosphate dibasic

puriss., meets analytical specification of NF, 99-102%

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

assay

99-102%

quality

meets analytical specification of NF

impurities

≤0.001% heavy metals (as Pb)

pH

7.6-8.2 (25 °C, 1%)

mp

155 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤2 mg/kg
Fe: ≤10 mg/kg

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sakakura et al.
Journal of chromatography, 621(2), 123-131 (1993-11-24)
A method for the simultaneous determination of bile acids in rat bile and serum by high-performance liquid chromatography with a post-column enzymic reaction and fluorescence detection has been developed. Without prior fractionation and alkaline hydrolysis, 26 unconjugated, glycine- and taurine-conjugated
Hydrothermal synthesis, structures and luminescent properties of nanocrystaline Ca8Gd2(PO4)6O2: Eu2+, Eu3+ phosphors.
Bharat L, et al.
Chemical Engineering Journal, 240, 179-186 (2014)
Electric Field-Assisted Solid Phase Extraction: Study of Electrochromatographic Parameters with an Anionic Model Compound.
Orlando RM, et al.
Journal of the Brazilian Chemical Society, 26(2), 310-318 (2015)
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of
Xiaopeng Gao et al.
Journal of the science of food and agriculture, 90(5), 750-758 (2010-04-01)
A 3-year field trial was conducted to investigate the effect of agricultural management practices including tillage, preceding crop and phosphate fertilization on root colonization by arbuscular mycorrhizal (AM) fungi and grain cadmium (Cd) concentration of durum wheat (Triticum turgidum L.).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service