02119
Sodium bromide
puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
About This Item
Recommended Products
vapor pressure
1 mmHg ( 806 °C)
grade
puriss.
assay
99-100.5% (calc. to the dried substance)
quality
meets analytical specification of Ph. Eur., BP, USP
impurities
acidity or alkalinity, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.02% Mg, alkaline earth metals (as Ca)
loss
≤1% loss on drying, 120 °C
pH
5-7 (20 °C, 5%)
mp
755 °C (lit.)
anion traces
bromate (BrO3-): ≤10 mg/kg
chloride (Cl-): ≤2000 mg/kg
iodide (I-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
As: ≤2 mg/kg
Ba: ≤20 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
Mg: ≤10 mg/kg
suitability
complies for appearance of solution
SMILES string
[Na+].[Br-]
InChI
1S/BrH.Na/h1H;/q;+1/p-1
InChI key
JHJLBTNAGRQEKS-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
- As catalyst for the microwave assisted three-component cyclocondensation of aryl aldehydes, alkyl nitriles and dimedone for the synthesis of highly functionalized tetrahydrobenzo[b]pyrans.[3]
- To compose the aqueous two-phase system (ATPS) containing Gemini surfactant ethanediyl-1,2-bis(dodecyldimethylammonium bromide).[1]
- Preparation of unoxidized and carboxylated pullulan, which are employed as mediators for the preparation of silver nanoparticles (AgNPs).[4]
signalword
Warning
hcodes
Hazard Classifications
Repr. 2 - STOT RE 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service