02110
Potassium bromide
puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5%
About This Item
Recommended Products
vapor pressure
<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)
grade
puriss.
assay
99.5-100.5%
quality
meets analytical specification of Ph. Eur., BP, USP
impurities
acidity or alkalinity, complies
residual solvents, complies
≤0.0005% ammonium (NH4)
≤0.001% heavy metals (as Pb)
≤0.02% Mg, alkaline earth metals (as Ca)
loss
≤0.3% loss on drying, 130 °C
pH
5-8 (20 °C, 5%)
mp
734 °C (lit.)
anion traces
bromate (BrO3-): ≤10 mg/kg
chloride (Cl-): ≤2000 mg/kg
iodide (I-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
As: ≤3 mg/kg
Ba: ≤10 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
Mg: ≤20 mg/kg
Na: ≤1500 mg/kg
suitability
complies for appearance of solution
SMILES string
[K+].[Br-]
InChI
1S/BrH.K/h1H;/q;+1/p-1
InChI key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service