Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z805890

Hellma® liquid calibration standards set

UV1/UV1H; UV10/UV10H; UV11/UV11H; UV19/UV19H, for testing the stray light acc. to USP 857

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.32

manufacturer/tradename

Hellma 667-UV105H

Looking for similar products? Visit Product Comparison Guide

General description

Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to stray light.

Consisting of :
Potassium chloride in purified water (667-UV1, wavelength [nm] 200 (cut-off), pathlength 10 mm
Potassium chloride in purified water, reference filter (667-UV1H, wavelength [nm] 200 (cut-off), pathlength 5 nm
Sodium iodide in purified water (667-UV10, wavelength [nm] 259 cut-off), pathlength 10 mm
Sodium iodide in purified water, reference filter (667-UV10H, wavelength [nm] 259 (cut-off), pathlength 5 mm
Sodium nitrite in purified water (667-UV11, wavelength [nm] 385 (cut-off), pathlength 10 mm
Sodium nitrite in purified water, reference filter (667-UV11H, wavelength [nm] 385 (cut-off), pathlength 5 mm
Acetone (667-UV19, wavelength [nm] 325 (cut-off), pathlength 10 mm
Acetone, reference filter (667-UV19H, wavelength [nm] 325 (cut-off), pathlength 5 mm

Legal Information

Hellma is a registered trademark of Hellma GmbH

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Ox. Liq. 3 - STOT RE 2 Oral - STOT SE 3

target_organs

Central nervous system, Thyroid

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service