Key Documents
Z804800
Hellma® liquid calibration standards set
UV5,UV25 for testing the wavelength accuracy acc. to USP 857
About This Item
UNSPSC Code:
12352200
manufacturer/tradename
Hellma 667-UV425
technique(s)
UV/Vis spectroscopy: suitable
General description
Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to wavelength accuracy.
Consisting of :
Holmium in perchloric acid (667-UV5, wavelength 241; 250; 278, 287; 333; 345; 361; 385; 416; 42; 468; 485; 536; 640 nm)
Didymium in perchloric acid (667-UV25, wavelength 732; 740; 794; 801; 864 nm)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)