Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z804797

Enhanced UV/VIS Spectrophotometer Kit

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

manufacturer/tradename

Jaytee Biosciences Ltd (CRM-2101)

technique(s)

UV/Vis spectroscopy: suitable

application(s)

food and beverages
petroleum
pharmaceutical
plastics

format

neat

General description

Contains qualification standards for
  • UV Photometric Accuracy
  • UV Resolution
  • UV Stray Light
  • UV Multi-Wavelength

Application

Enhanced UV/VIS Spectrophotometer Kit for use in the qualification of UV spectrophotometers.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory as per UKAS (United Kingdom Accreditation Service)
  • Supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification
  • Traceable instrument qualification adds confidence to all data generated
  • Accompanied with a certificate of analysis (COA) having information on the expiry date for an effective audit

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Thyroid

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

71.6 °F

flash_point_c

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service