Key Documents
Z804681
Hellma® liquid calibration standards set
UV5,UV14 for testing the wavelength accuracy
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
Hellma 667-UV400
technique(s)
UV/Vis spectroscopy: suitable
General description
Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to wavelength accuracy.
Consisting of :
Holmium in perchloric acid (667-UV5, wavelength 241; 287; 361; 536; 640 nm)
Perchloric acid reference filter (667-UV14, wavelength 235; 257; 313; 350 nm)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)