Key Documents
Z804673
Specac® Pearl Liquid Analyser Oyster bottom
CaF2, pathlength 1000 μm, wedged windows
About This Item
UNSPSC Code:
12352200
material
CaF2
feature
wedged windows
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
pathlength
1000 μm
General description
The Pearl is a high specification liquid transmission accessory designed for laboratory spectroscopic sample analysis in the Near and Mid-Infrared. The accessory contains Specac′s innovative Oyster Cell assembly which holds your sample in a horizontal plane.
The design of the Pearl and Oyster Cell allows for fast and easy application of the sample, and for cleaning between samples.
A complete Oyster liquid cell consists of a separate top and bottom window assembly which are brought together to create a defined, consistent and repeatable pathlength for the liquid cell for the containment of a liquid sample between the inner contacting window surfaces.
The Oyster cell can be provided as two different types (parallel or wedged angle) of a complete liquid cell assembly fitted with either ZnSe or CaF2 window material options.
The Oyster top window assembly fitted with a particular window material is common to all of the complete Oyster cells offered.
The Oyster bottom window assembly is offered as a choice for the fitting of ZnSe or CaF2 window material at a parallel or wedge angle window setting. A pathlength option to choose is from the 25, 50, 100, 200, 500 and 1000 um offerings.
Features and Benefits
  • The easiest to use FTIR liquid sampling system available Faster, more accurate and more repeatable than traditional liquid cells
  • Wedged cell option to eliminate troublesome fringing completely
  • Different pathlengths and window materials changed in seconds
  • Handles viscous materials with ease
Linkage
A Specac® Benchmark baseplate for your specific spectrometer is needed for installing the Pearl Liquid Transmission Accessory. A Pearl optical unit Z805432 is required plus a complete Oyster of the window type, pathlength and wedging.
Legal Information
Oyster is a trademark of Specac, Inc.
Pearl is a trademark of Specac, Inc.
Specac is a registered trademark of Specac Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)