Z804665
UV Stray Light Standards
certified reference material
About This Item
Recommended Products
grade
certified reference material
manufacturer/tradename
Jaytee Biosciences Ltd (CRM-2001)
technique(s)
UV/Vis spectroscopy: suitable
General description
These UV Stray Light Standards are designed to qualify the Stray Light of UV Spectrophotometers to the limits defined in the EP.
Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.
All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.
Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.
Application
Features and Benefits
- Product is of certified reference material grade
- An ampoule of standard and an ampoule of blank are included
- CoA has the instruction on how to qualify the instrument
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service