Z804568
UV Resolution Standards
certified reference material
About This Item
Recommended Products
grade
certified reference material
manufacturer/tradename
Jaytee Biosciences Ltd (CRM-2002)
technique(s)
UV/Vis spectroscopy: suitable
General description
These UV Resolution Standards are used to qualify the UV Resolution of UV Spectrophotometers to the limits defined in the EP.
Developed to be used in any laboratory regardless of the regulator and are as relevant to a pharmaceutical company as to a contract lab working to ISO 17025.
All products come supplied in flame-sealed ampoules to increase the shelf life and prevent contamination, with full certification.
Traceable instrument qualification adds confidence to all data generated. In addition, your audit trail is backed up by the independent verification by UKAS to ISO 17034.
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3
target_organs
Central nervous system, Nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
71.6 °F
flash_point_c
22 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service