Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z804355

Hellma® liquid calibration standards

UV25, for testing the wavelength accuracy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.32

manufacturer/tradename

Hellma 667-UV25

technique(s)

UV/Vis spectroscopy: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to wavelength accuracy

The Diymium filter consists of a solution of Praseodymium and Neodymium dissolved in perchloric acid. This filter is highly suitable for checking the wavelength accuracy of spectrophotometers both in the UV and the visible range. In the range between 329 nm and 864 nm the filter shows a spectrum with a multitude of highly defined and characteristic peaks. .

Consisting of :
Didymium in perchloric acid (667-UV25, wavelength 329; 469; 575; 740, 864 nm)

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Thyroid

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service