Z804029
Hellma® calibration standard, glass filter set
F390, F301, F303, for testing the wavelength accuracy and the photometric accuracy
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
Hellma 666-S300
technique(s)
UV/Vis spectroscopy: suitable
General description
Secondary calibration standard for wavelength accuracy and photometric accuracy. Certified.
Glass Filter Set: F390, F301, F303, F0 (nominal absorbance: 0.04; 2.5; 3.0 at 440; 465; 546,1; 590; 635 nm)
Glass Filter Set: F390, F301, F303, F0 (nominal absorbance: 0.04; 2.5; 3.0 at 440; 465; 546,1; 590; 635 nm)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)