Key Documents
Z803928
Hellma® calibration standard, glass filter set
F201, F202, F203, F0, for testing the wavelength accuracy and the photometric accuracy
About This Item
UNSPSC Code:
12352200
manufacturer/tradename
Hellma 666-S004
technique(s)
UV/Vis spectroscopy: suitable
General description
Secondary calibration standard for wavelength accuracy and photometric accuracy. Certified.
Glass Filter Set: F201, F202, F203, F0 (peak positions in nm, A: 440; 465; 546,1; 590; 635)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)