Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z802654

Hellma® liquid calibration standards set

UV60, UV600, UV14, for testing the photometric accuracy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
SB.32

grade

certified reference material

manufacturer/tradename

Hellma 667-UV305

application(s)

food and beverages
petroleum
pharmaceutical
plastics

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Consisting of :
60 mg Potassium dichromate in perchloric acid (667-UV60, wavelength 235; 257; 313; 350 nm)
600 mg Potassium dichromate in perchloric acid (667-UV600, wavelength 430 nm)
Perchloric acid reference filter (667-UV14, wavelength 235; 257; 313; 350 nm)

Potassium dichromate dissolved in perchloric acid is a highly suitable method to check for photometric accuracy (absorbance) in the UV region. Potassium dichromate gives a spectral scan containing characteristic peak maxima at 257 nm and 350 nm and minima at 235 nm and 313 nm. The potassium dichromate solution is sealed in an airtight sealed quartz precision cell with 10 mm light path. The absorbance values of the filter are determined at the wavelengths 235 nm, 257 nm, 313 nm, and 350 nm and are specified in the certificate. It should be noted that the measurements are carried out with the filter 667-UV14 (0.01 N perchloric acid) in the reference beam path. Measured values of the reference filter are separately displayed in the certificate.

Application

Hellma® liquid calibration standards set for checking the photometric accuracy of spectrophotometers in accordance with European Pharmacopoeia (Ph. Eur.) and U.S. Pharmacopeia (USP) 857.

Features and Benefits

  • Accredited to DAkkS Deutsche Akkreditierungsstelle (National accreditation body for the Federal Republic of Germany)
  • Traceable to NIST (National Institute of Standards and Technology)
  • Assured Quality of the measurement results
  • Proven accuracy and reliability

Legal Information

Hellma is a registered trademark of Hellma GmbH

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service