Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z802530

Hellma® liquid calibration standards set

UV6, UV9, for testing the spectral resolution acc. to Ph. Eur.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
SB.32

manufacturer/tradename

Hellma 667.200-UV

Looking for similar products? Visit Product Comparison Guide

General description

Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to resolution of the spectral bandwidth.

Consisting of :
Toluene in n-Hexane (667-UV6, scan 265 - 270 nm)
n-Hexane reference filter (667-UV9, scan 265 - 270 nm)

To check the resolution power of a spectrophotometer, the ratio of the peak maximum at λmax (269 nm) to the absorbance of the minimum at λmin (266 nm) is calculated. This gives a measure of the spectral bandwidth of the instrument.

Determination of the resolution is carried out against the 667-UV9 hexane blank reference filter, which is supplied with every filter set. Measurement is the same for all bandwidths. The wavelength range between 265 nm and 270 nm is scanned and the peak maxima and minima are determined. Then the peak/trough ratio is calculated and compared to the reference values.

Legal Information

Hellma is a registered trademark of Hellma GmbH

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service