Z802476
Hellma® liquid calibration standards set
UV11, UV12, for testing the stray light
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151816
NACRES:
SB.32
manufacturer/tradename
Hellma (667-UV102)
Looking for similar products? Visit Product Comparison Guide
General description
Certified liquid Hellma calibration standard to enable the checking of spectrophotometers with respect to stray light.
Consisting of :
Sodium nitrite in pure water (667-UV11, wavelength [nm] 385 (cut-off))
Pure water reference filter (667-UV12, wavelength 198; 200; 300; 400 nm)
Consisting of :
Sodium nitrite in pure water (667-UV11, wavelength [nm] 385 (cut-off))
Pure water reference filter (667-UV12, wavelength 198; 200; 300; 400 nm)
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
signalword
Warning
hcodes
Hazard Classifications
Ox. Liq. 3
Storage Class
5.1B - Oxidizing hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service