Key Documents
Z802093
Hellma® empty filter mount for calibration standards
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
manufacturer/tradename
Hellma 666-F0
application(s)
food and beverages
petroleum
pharmaceutical
plastics
Application
Hellma® empty filter mount for calibration standards as a reference filter frame made of aluminum.
Features and Benefits
  • Accredited to DAkkS (Deutsche Kalibrierdienst-German Calibration Authority)
  • Manufactured in calibration laboratory
  • Assured Quality
  • Proven accuracy and reliability
  • Fast and reliable recertification service
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)