Key Documents
Z801771
Specac® sealed flow cell for IR spectroscopy
CaF2 windows, 0.1 mm pathlength
About This Item
UNSPSC Code:
23151816
NACRES:
SB.32
General description
Sealed IR transmission cell with 1/16" Swagelok fittings for flow measurements. Can be used in Specac Ambient Temperature Holders at ambient temperatures or in suitable heating jackets.
Legal Information
Specac is a registered trademark of Specac Ltd.
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)