Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z801429

Hellma® absorption cuvettes, semi Micro

Suprasil® quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 1,400 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.32

material

Suprasil® quartz

description

2 polished windows

manufacturer/tradename

Hellma 117.104-QS

parameter

200-2,500 nm spectral range

H × W × D

56 mm × 12.5 mm × 12.5 mm

chamber volume

1,400 μL

pathlength

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

with septum screw cap

Packaging

Sold as packages containing a single cell (1EA = 1 cell)

Legal Information

Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service