Z801364
Hellma® calibration standard, Didymium glass filter
F7A, for testing the photometric accuracy
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
Recommended Products
manufacturer/tradename
Hellma (666-F7A)
General description
Secondary calibration standard for the photometric accuracy. Certified.
Didymium Glass Filter F7A nominal value of the absorption ~0.5-1.0 (peak positions in nm, A: 270; 280; 297; 320; 340)
Didymium Glass Filter F7A nominal value of the absorption ~0.5-1.0 (peak positions in nm, A: 270; 280; 297; 320; 340)
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)