Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z800198

Hellma® liquid calibration standards

UV11, for testing the stray light

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

manufacturer/tradename

Hellma 667-UV11

technique(s)

UV/Vis spectroscopy: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to stray light.

Consisting of :
Sodium nitrite in purified water (667-UV11, wavelength [nm] 385 (cut-off), pathlength 10 mm

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

pictograms

Flame over circle

signalword

Warning

hcodes

Hazard Classifications

Ox. Liq. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service