Key Documents
Z800007
LICO 500 - Spectral colorimeter
About This Item
UNSPSC Code:
41115710
manufacturer/tradename
Hach Lange (LMV185.99.00001)
General description
Included accessories:
  • Universal power supply 100-240V, 50-60Hz, with exchangeable plug adapters for EU, GB, US, China
  • Dust cover
  • User Manual:
Z800007EN-1EA : English + German manual
Z800007IT-1EA : plus Italian manual
Z800007FR-1EA : plus French manual
Features and Benefits
High performance, microprocessor controlled spectrophotometer with a wavelengths range from 380 to 720 nm for color measurement or 320 nm up to 1100 nm for routine analyses. The LICO 500 can carry out an exact colorimetric evaluation in conformity with several ISO/ASTM standards with just a single measurement and display the result in terms of traditional color systems such as Iodine, Hazen/APHA or Gardner color numbers as well as in modern CIE-L*a*b* color values. Besides the over 20 color indexes, transmittance and absorbance can be measured at individual wavelengths, so that the LICO 500 can also be used universally for analytical purposes in the laboratory. Touch- screen TFT-Color-Display, Automatic cuvette recognition, Data log for 500 color values, 50 color reference values, 500 photometric readings, 20 wavelength scans, 20 time scans, Automatic zero calibration program, Reference Beam Technology, Password protection, GLP documentation, USB-Ports 1 x Type A and 1 x Type B Color measurement methods : Iodine, Hazen/APHA, Gardner-color Saybolt, Mineral oil, Klett-color Hess-Ives, ADMI, Yellowness-index AOCS- Red/Yellow, Chlorophyll A CIE- Lab, Hunter-Lab, XYZ, European and US Pharmacopoeia
Choose from one of the most recent versions:
Certificates of Analysis (COA)