Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001128

Epinastine hydrochloride for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Epinastine hydrochloride, 9,13b-Dihydro-1H-dibenz[cf]imidazo[1,5-a]azepine hydrochloride, WAL-801Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3 · HCl
CAS Number:
Molecular Weight:
285.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].NC1=NCC2N1c3ccccc3Cc4ccccc24

InChI

1S/C16H15N3.ClH/c17-16-18-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)19(15)16;/h1-8,15H,9-10H2,(H2,17,18);1H

InChI key

VKXSGUIOOQPGAF-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Epinastine hydrochloride is a non-sedating H1 histamine receptor antagonist.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Lambiase et al.
Current opinion in allergy and clinical immunology, 9(5), 454-465 (2009-08-05)
Multiple action drugs, such as azelastine, epinastine, ketotifen and olopatadine, have recently been suggested to combine antihistaminic effect, mast cell stabilization and anti-inflammatory action. This pharmaceutical class is, therefore, rapidly becoming the first choice for prevention and treatment for allergic
Takeo Tsujii et al.
Psychopharmacology, 207(1), 127-132 (2009-08-26)
Antagonists of histamine H(1) receptors (antihistamines) are widely used for the treatment of allergic disorders in children. These drugs' sedative effect on brain function, however, has been mostly examined in adults. The objective of this study was to examine the
Yukihisa Matsumoto et al.
Neurobiology of learning and memory, 101, 103-113 (2013-02-06)
Sensory preconditioning (SPC) is a procedure to demonstrate learning to associate between relatively neutral sensory stimuli in the absence of an external reinforcing stimulus, the underlying neural mechanisms of which have remained obscure. We address basic questions about neural processes
Hiroyuki Mizuguchi et al.
Journal of pharmacological sciences, 108(4), 480-486 (2008-12-17)
Antihistamines are effective for treatment of seasonal nasal allergy. Recently, prophylactic treatment with antihistamines in patients with pollinosis was reported to be more effective when started before the pollen season. The administration with antihistamines from 2 to 6 weeks before
Masanori Ito et al.
International journal of pharmaceutics, 441(1-2), 121-127 (2012-12-19)
The study objective was to quantitatively predict a drug's bitterness and estimate bitterness masking efficiency using an electronic tongue (e-Tongue). To verify the predicted bitterness by e-Tongue, actual bitterness scores were determined by human sensory testing. In the first study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service