Y0001011
Spanish sage oil for peak identification
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Sage oil
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
spanish sage oil
manufacturer/tradename
EDQM
refractive index
n20/D 1.468 (lit.)
bp
181 °C (lit.)
density
0.912 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
Storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
116.6 - 129.2 °F
flash_point_c
47 - 54 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service