Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000715

Oxitropium bromide impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Scopolamine methyl bromide, Hyoscine methyl bromide, Methscopolamine bromide

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$118.00

$118.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$118.00

About This Item

Empirical Formula (Hill Notation):
C18H24NO4 · Br
CAS Number:
Molecular Weight:
398.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$118.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

oxitropium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].[H][C@]12C[C@@H](C[C@]([H])([C@@]3([H])O[C@@]13[H])[N+]2(C)C)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C18H24NO4.BrH/c1-19(2)14-8-12(9-15(19)17-16(14)23-17)22-18(21)13(10-20)11-6-4-3-5-7-11;/h3-7,12-17,20H,8-10H2,1-2H3;1H/q+1;/p-1/t12-,13-,14-,15+,16-,17+;/m1./s1

InChI key

CXYRUNPLKGGUJF-RAFJPFSSSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

application

Oxitropium bromide impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramona Schrage et al.
Biochemical pharmacology, 90(3), 307-319 (2014-05-28)
Activation of G protein-coupled receptors involves major conformational changes of the receptor protein ranging from the extracellular transmitter binding site to the intracellular G protein binding surface. GPCRs such as the muscarinic acetylcholine receptors are commonly probed with radioantagonists rather

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service