Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000251

Molgramostim

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 0.3 mL. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

PLEASE NOTE: TEMPORARILY UNAVAILABLE DUE TO RESTRICTED QUANTITY

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Méry et al.
Dermatology (Basel, Switzerland), 208(2), 135-137 (2004-04-02)
Leg ulcers are a frequently neglected, severe complication of sickle cell disease (SCD), responsible for significant altering quality of life. The management of leg ulcers remains disappointing. Local application of recombinant human granulocyte-macrophage colony-stimulating factor has been shown to be
Chen Chen et al.
Journal of Asian natural products research, 14(9), 877-885 (2012-08-29)
The antivirus effect of quercetin and oseltamivir on the Toll-like receptor 7 (TLR7) signaling pathway was observed when dendritic cells and macrophages were infected with H1N1. Leukomonocytes were obtained from umbilical cord blood and harvested after stimulation by recombinant human
Maria Liljefors et al.
Cancer immunology, immunotherapy : CII, 57(3), 379-388 (2007-08-07)
Recombinant granulocyte-macrophage colony-stimulating factor (GM-CSF) is used in immunotherapy for correction of neutropoenia. The optimal dose for activation of immune functions and the pharmacokinetics following repeated administrations is less analysed in depth. In this study, the pharmacokinetics and the effects
Guglielmo Mario Actis Dato et al.
The International journal of artificial organs, 35(1), 67-76 (2012-01-31)
Coronary artery disease (CAD), along with its main complications such as acute myocardial infarction (AMI) and congestive heart failure (CHF), remains a serious worldwide problem and affects many patients despite the improvement of medical treatment. The possibility of the replacement
X Thomas et al.
Leukemia, 21(3), 453-461 (2007-01-26)
In a multicenter trial, 259 young adults (15-49 years) with newly diagnosed acute myeloid leukemia (AML) were first randomized to receive a timed-sequential induction regimen given either alone (135 patients) or concomitantly with granulocyte-macrophage colony-stimulating factor (GM-CSF) (124 patients). Patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service