Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

XRFOX44

Supelco

Silica XRF Standard - >99.99%

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

manufacturer/tradename

RTC XRF-OX-44

technique(s)

XRF: suitable

matrix

silicon dioxide

application(s)

environmental

format

matrix material

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service