Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VT000325

Supelco

Staphylococcus aureus subsp. aureus WDCM 00032 Vitroids

600-1’400 CFU mean value range, certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

form

disc (hydrosoluble)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Staphylococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Vitroids discs contain Staphylococcus aureus subsp. aureus WDCM 00032 viable microorganisms (600-1′400 CFU) in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids are derived from a traceable culture obtained freeze-dried from CECT®.

Application

  • Daily quality control
  • Performance testing of media acc. ISO 11133
  • Validation of new methods
  • Materials for proficiency testing or ring trials
  • Method development
  • Staff training
  • Starter cultures

Features and Benefits

  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use

Other Notes

ATCC® 6538, NCTC 10788, NCPF -, DSM 799, LMG 8064, IHEM -, CIP 4.83
Origin Strain No. CECT® 239

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service