Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VT000265

Supelco

Pseudomonas paraeruginosa WDCM 00026 Vitroids

600-1’400 CFU mean value range, certified reference material, suitable for microbiology

Synonym(s):

Microbiological standards, Certified reference material, P. aeruginosa WDCM 00026 Vitroids, Microbiological standards, Certified reference material, P. paraeruginosa WDCM 00026 Vitroids, Pseudomonas aeruginosa WDCM 00026 Vitroids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

form

disc (hydrosoluble)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Pseudomonas spp.

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Other Notes

ATCC® 9027, NCTC 12924, NCPF -, DSM 1128, LMG 8029, IHEM -, CIP 82.118
Origin Strain No. CECT® 111

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service